Invitrogen
Manufacturer Code:MA527653
Catalog # PIMA527653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S43-6 |
Immunogen | Fusion protein amino acids 2-77 of human KCNQ4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DFNA 2 DFNA2 KCNQ 4 Kcnq4 KCNQ4&lowbarHUMAN KQT like 4 KQT-like 4 KV7.4 Potassium channel KQT like 4 Potassium channel subunit alpha KvLQT4 Potassium voltage gated channel KQT like protein 4 Potassium voltage gated channel KQT like subfamily member 4 Potassium voltage gated channel subfamily KQT member 4 Potassium voltage-gated channel subfamily KQT member 4 Voltage gated potassium channel subunit Kv7.4 Voltage-gated potassium channel subunit Kv7.4; KQT-like 4; potassium channel KQT-like 4; Potassium channel subunit alpha KvLQT4; potassium channel, voltage gated KQT-like subfamily Q, member 4; Potassium voltage-gated channel subfamily KQT member 4; potassium voltage-gated channel, KQT-like subfamily, member 4; potassium voltage-gated channel, subfamily Q, member 4; Voltage-gated potassium channel subunit Kv7.4
Gene Aliases: DFNA2; DFNA2A; KCNQ4; KV7.4
UniProt ID: (Human) P56696, (Mouse) Q9JK97
Entrez Gene ID: (Human) 9132, (Mouse) 60613, (Rat) 298496
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.