Abnova
Manufacturer Code:MAB6656
Catalog # 89-112-982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S43-6 |
Immunogen | Recombinant fusion protein corresponding to amino acids 2-77 of human KCNQ4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant KCNQ4.
For Research Use Only
Protein Aliases: DFNA2; KQT-like 4; KV7.4; potassium channel KQT-like 4; Potassium channel subunit alpha KvLQT4; potassium channel, voltage gated KQT-like subfamily Q, member 4; Potassium voltage-gated channel subfamily KQT member 4; potassium voltage-gated channel, KQT-like subfamily, member 4; Voltage-gated potassium channel subunit Kv7.4
Gene Aliases: DFNA2; DFNA2A; KCNQ4; KV7.4
UniProt ID: (Human) P56696
Entrez Gene ID: (Human) 9132
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.