Invitrogen
Manufacturer Code:PA541020
Catalog # PIPA541020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KCNQ4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KQT-like 4; potassium channel KQT-like 4; Potassium channel subunit alpha KvLQT4; potassium channel, voltage gated KQT-like subfamily Q, member 4; potassium voltage-gated channel KQT-like protein 4; Potassium voltage-gated channel KQT-like subfamily member 4 DFNA2 KV7.4 DFNA2A; Potassium voltage-gated channel subfamily KQT member 4; potassium voltage-gated channel, KQT-like subfamily, member 4; potassium voltage-gated channel, subfamily Q, member 4; Voltage-gated potassium channel subunit Kv7.4
Gene Aliases: DFNA2; DFNA2A; KCNQ4; KV7.4
UniProt ID: (Human) P56696, (Mouse) Q9JK97
Entrez Gene ID: (Dog) 482451, (Human) 9132, (Rabbit) 100358271, (Pig) 100526228, (Horse) 100067963, (Guinea pig) 100716024, (Mouse) 60613, (Rat) 298496
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.