Invitrogen
Manufacturer Code:PA595399
Catalog # PIPA595399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human KCNQ1 different from the related mouse sequence by two amino acids and from the related rat sequence by one amino acid. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1; kidney and cardiac voltage dependend K+ channel; KQT-like 1; potassium channel, voltage gated KQT-like subfamily Q, member 1; Potassium voltage-gated channel subfamily KQT member 1; Potassium voltage-gated channel subfamily KQT member 1 IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1 KQT-like 1 Voltage-gated potassium channel subunit Kv7.1 KCNQ1 KCNA8 KCNA9 KVLQT1; potassium voltage-gated channel, KQT-like subfamily, member 1; potassium voltage-gated channel, subfamily Q, member 1; slow delayed rectifier channel subunit; Voltage-gated potassium channel subunit Kv7.1
Gene Aliases: ATFB1; ATFB3; JLNS1; KCNA8; KCNA9; KCNQ1; Kv1.9; Kv7.1; KVLQT1; LQT; LQT1; RWS; SQT2; WRS
UniProt ID: (Human) P51787, (Rat) Q9Z0N7
Entrez Gene ID: (Human) 3784, (Rat) 84020
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.