Invitrogen
Manufacturer Code:PA519093
Catalog # PIPA519093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EQLTVPRRGPDEGS) corresponding to the C-terminus amino acids of KCNQ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1; KCNQ1 potassium voltage-gated channel KQT-like subfamily member 1 ATFB1 FLJ26167 JLNS1 KCNA8 KCNA9 KVLQT1 Kv1.9 Kv7.1 LQT LQT1 RWS SQT2 WRS Jervell and Lange-Nielsen syndrome 1 kidney and cardiac voltage dependend K+ channel long (electrocardiographic) QT syndrome Ward-Romano syndrome 1 slow delayed rectifier channel subunit; kidney and cardiac voltage dependend K+ channel; KQT-like 1; potassium channel, voltage gated KQT-like subfamily Q, member 1; Potassium voltage-gated channel subfamily KQT member 1; potassium voltage-gated channel, KQT-like subfamily, member 1; slow delayed rectifier channel subunit; Voltage-gated potassium channel subunit Kv7.1
Gene Aliases: ATFB1; ATFB3; JLNS1; KCNA8; KCNA9; KCNQ1; Kv1.9; Kv7.1; KVLQT1; LQT; LQT1; RWS; SQT2; WRS
UniProt ID: (Human) P51787
Entrez Gene ID: (Human) 3784
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.