Invitrogen
Manufacturer Code:PA577620
Catalog # PIPA577620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KQIGSLESKLEHLTAS corresponding to amino acid residues 659-674 of human KCa 2.3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KCa2.3; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 3; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 3; SK3; SK3 KCNN3; SKCa 3; SKCa3; small conductance calcium-activated potassium channel 3; Small conductance calcium-activated potassium channel protein 3; small-conductance Ca2+-activated K+ channel 3
Gene Aliases: hSK3; K3; KCa2.3; KCNN3; SK3; SKCA3
UniProt ID: (Human) Q9UGI6, (Mouse) P58391, (Rat) P70605
Entrez Gene ID: (Human) 3782, (Mouse) 140493, (Rat) 54263
Molecular Function: ion channel potassium channel transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.