Invitrogen
Manufacturer Code:PA541071
Catalog # PIPA541071
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KCNN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apamin-sensitive small-conductance Ca2+-activated potassium channel; KCa2.2; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 2; Potassium intermediate/small conductance Calcium-activated channel subfamily N member 2 KCa2.2 SK2 SKCA2 hSK2; potassium intermediate/small conductance calcium-activated channel type2; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 2; SK2; SKCa 2; SKCa2; small conductance calcium-activated channel, subfamily N, member 2; small conductance calcium-activated potassium channel 2; Small conductance calcium-activated potassium channel protein 2; small conductance calcium-activated potassium channel protein 2-like; small conductance calcium-activated potassium channel subfamily N member 2; small conductance potassium channel type 2
Gene Aliases: fri; hSK2; KCa2.2; KCNN2; SK2; SKCa 2; SKCA2
UniProt ID: (Human) Q9H2S1, (Rat) P70604, (Mouse) P58390
Entrez Gene ID: (Rabbit) 100342605, (Bovine) 404177, (Human) 3781, (Horse) 100064342, (Dog) 474640, (Guinea pig) 100734732, (Rat) 54262, (Mouse) 140492
Molecular Function:
ion channel
potassium channel
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.