Novus Biologicals
Manufacturer Code:NBP2841130.1ML
Catalog # NB032041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of mouse KCNN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apamin-sensitive small-conductance Ca2+-activated potassium channel; hSK2 KCa2.2SKCA2 potassium intermediate/small conductance calcium-activated channel subfamilyN member 2 SK2apamin-sensitive small-conductance Ca2+-activated potassium channel SKCa 2 SKCa2 small conductance calcium-activated potassium channel protein 2; KCa2.2; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 2; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 2; SK2; small conductance calcium-activated potassium channel 2; Small conductance calcium-activated potassium channel protein 2
Gene Aliases: hSK2; KCa2.2; KCNN2; SK2; SKCa 2; SKCA2
UniProt ID: (Human) Q9H2S1
Entrez Gene ID: (Human) 3781
Molecular Function:
ion channel
potassium channel
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.