Invitrogen
Manufacturer Code:PA577600
Catalog # PIPA577600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DRPGSGKPPTVSHRLGHRR corresponding to amino acid residues 75-93 of rat KCa 2.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KCa2.1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1-like; SK1; SK1 KCNN1; SKCa 1; SKCa1; Small conductance calcium-activated potassium channel protein 1
Gene Aliases: AI854248; KCa2.1; Kcnn1; SK1
UniProt ID: (Rat) P70606, (Mouse) Q9EQR3
Entrez Gene ID: (Rat) 54261, (Mouse) 84036
Molecular Function:
ion channel
potassium channel
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.