Novus Biologicals
Manufacturer Code:NBP30481120UL
Catalog # NB095141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human KCNN1 (NP_002239.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSK1 KCa2.1 potassium intermediate/small conductance calcium-activated channel subfamily N member 1 SK SK1 SKCa 1 SKCA1 small conductance calcium-activated potassium channel protein 1; KCa2.1; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1; SK1; small conductance calcium-activated potassium channel 1; Small conductance calcium-activated potassium channel protein 1
Gene Aliases: hSK1; KCa2.1; KCNN1; SK; SK1; SKCA1
UniProt ID: (Human) Q92952
Entrez Gene ID: (Human) 3780
Molecular Function: ion channel potassium channel transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.