Novus Biologicals
Manufacturer Code:NBP212917
Catalog # NBP212917
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S53-32 |
Immunogen | Fusion protein amino acids 1-41 (N-terminus) and 218-235 (C-terminus) of mouse BKBeta2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: big potassium channel beta subunit 2; BK channel beta subunit 2; BK channel subunit beta-2; BK channel subunit beta-2 BKbeta2 calcium-activated potassium channel subunit beta-2 Calcium-activated potassium channel subfamily M subunit beta-2 Charybdotoxin receptor subunit beta-2 hbeta2 Hbeta3 k(VCA)beta-2 large conductance calcium-activated potassium channel beta 2 subunit large-conductance Ca2+-activated K+ channel beta2 subunit Maxi K channel subunit beta-2 MaxiK channel beta 2 subunit MGC22431 potassium large conductance calcium-activated channel subfamily M beta member2 slo-beta-2; BKbeta2; Calcium-activated potassium channel subunit beta-2; Calcium-activated potassium channel, subfamily M subunit beta-2; Charybdotoxin receptor subunit beta-2; hbeta2; Hbeta3; hCG1646471; K(VCA)beta-2; large conductance calcium-activated potassium channel beta 2 subunit; large-conductance Ca2+-activated K+ channel beta2 subunit; Maxi K channel subunit beta-2; MaxiK channel beta 2 subunit; MaxiK channel beta-subunit 2; potassium channel subfamily M regulatory beta subunit 2; potassium large conductance calcium-activated channel, subfamily M, beta member 2; Slo-beta-2
Gene Aliases: KCNMB2
UniProt ID: (Human) Q9Y691
Entrez Gene ID: (Human) 10242
Molecular Function: ion channel potassium channel transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.