Invitrogen
Manufacturer Code:PA539067
Catalog # PIPA539067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human KCNMB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: big potassium channel beta subunit 2; BK channel beta subunit 2; BK channel subunit beta-2; BKbeta2; calcium-activated potassium channel beta 2; Calcium-activated potassium channel subunit beta-2; Calcium-activated potassium channel, subfamily M subunit beta-2; Charybdotoxin receptor subunit beta-2; hbeta2; Hbeta3; hCG1646471; K(VCA)beta-2; large conductance calcium-activated potassium channel beta 2 subunit; large-conductance Ca2+-activated K+ channel beta2 subunit; Maxi K channel subunit beta-2; MaxiK channel beta 2 subunit; MaxiK channel beta-subunit 2; potassium channel subfamily M regulatory beta subunit 2; potassium large conductance calcium-activated channel, subfamily M, beta member 2; Rbeta3; Slo-beta-2
Gene Aliases: 2700049B16Rik; 3110031N04Rik; Kcmb2; KCNMB2
UniProt ID: (Human) Q9Y691, (Mouse) Q9CZM9, (Rat) Q811Q0
Entrez Gene ID: (Human) 10242, (Mouse) 72413, (Rat) 294961
Molecular Function: ion channel potassium channel transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.