Invitrogen
Manufacturer Code:PA528284
Catalog # PIPA528284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 191 of Human KCNMB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: big potassium channel beta subunit 1; BK channel beta subunit; BK channel beta subunit 1; BK channel subunit beta-1; BKbeta; Calcium-activated potassium channel subunit beta; Calcium-activated potassium channel subunit beta-1; Calcium-activated potassium channel, subfamily M subunit beta-1; Charybdotoxin receptor subunit beta-1; K(VCA)beta SLO-BETA hslo-beta BK channel beta subunit BK channel subunit beta-1 BKbeta BKbeta1 Maxi K channel beta subunit calcium-activated potassium channel subunit beta calcium-activated potassium channel subunit beta-1 calcium-activated potassium channel subfamily M subunit beta-; K(VCA)beta-1; large conductance Ca2+-activated K+ channel beta 1 subunit; Maxi K channel subunit beta-1; MaxiK channel beta-subunit 1; potassium channel subfamily M regulatory beta subunit 1; potassium large conductance calcium-activated channel, subfamily M, beta member 1; Slo-beta; Slo-beta-1
Gene Aliases: BKbeta1; hbeta1; hslo-beta; K(VCA)beta; k(VCA)beta-1; KCNMB1; SLO-BETA; slo-beta-1
UniProt ID: (Human) Q16558, (Mouse) Q8CAE3
Entrez Gene ID: (Human) 3779, (Mouse) 16533
Molecular Function: ion channel potassium channel transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.