Invitrogen
Manufacturer Code:PA5114350
Catalog # PIPA5114350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide within rat KCNK9 aa 30-100. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid-sensitive potassium channel protein TASK-3; acid-sensitive potassium channel protein TASK-3 K2p9.1 Kcnk9 KT3.2 mgC138268 mgC138270 Potassium channel subfamily K member 9 potassium channel subfamily K member 9 (Task-3) potassium channel subfamily K member 9 potassium channel subfamily K member 9 (Task-3) potassium channel two pore domain subfamily K member 9 potassium two pore domain channel subfamily K member 9 TASK3 TASK-3 TWIK-related acid-sensitive K(+) channel 3 two pore K(+) channel KT3.2 Two pore potassium channel KT3.2; Potassium channel subfamily K member 9; potassium channel subfamily K member 9 (Task-3); potassium channel, subfamily K, member 9 (Task-3); potassium channel, two pore domain subfamily K, member 9; TWIK-related acid-sensitive K(+) channel 3; Two pore K(+) channel KT3.2; Two pore potassium channel KT3.2
Gene Aliases: K2p9.1; KCNK9; KT3.2; TASK-3; TASK3
UniProt ID: (Human) Q9NPC2, (Rat) Q9ES08
Entrez Gene ID: (Human) 51305, (Rat) 84429
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.