Novus Biologicals
Manufacturer Code:NBP146493
Catalog # NBP146493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the c-terminal region of mouse KCNK9 (TASK3) conjugated to an immunogenic carrier protein was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid-sensitive potassium channel protein TASK-3; KT3.2 potassium channel subfamily K member 9 TASK-3; Potassium channel subfamily K member 9; potassium channel, two pore domain subfamily K, member 9; TWIK-related acid-sensitive K(+) channel 3; Two pore K(+) channel KT3.2; Two pore potassium channel KT3.2
Gene Aliases: K2p9.1; KCNK9; KT3.2; TASK-3; TASK3
UniProt ID: (Human) Q9NPC2
Entrez Gene ID: (Human) 51305
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.