Invitrogen
Manufacturer Code:PA5106703
Catalog # PIPA5106703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KCNK7 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K2p7.1 KCNK7 potassium channel subfamily K member 7 potassium channel subfamily K member 7 potassium channel two pore domain subfamily K member 7 potassium two pore domain channel subfamily K member 7 RGD1565025 TWIK3 two pore domain K+ channel; Potassium channel subfamily K member 7; potassium channel, subfamily K, member 7; potassium channel, two pore domain subfamily K, member 7; two pore domain K+ channel
Gene Aliases: K2p7.1; KCNK7; RGD1565025; TWIK3
UniProt ID: (Human) Q9Y2U2
Entrez Gene ID: (Human) 10089, (Rat) 499303
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.