Novus Biologicals
Manufacturer Code:NBP2851350.1ML
Catalog # NB033064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human KCNK5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid-sensitive potassium channel protein TASK-2; Acid-sensitive potassium channel protein TASK-2 FLJ11035 K2p5.1 potassium channel subfamily K member 5 potassium channel subfamily K member 1 (TASK-2) potassium channel subfamily K member 5 TASK-2 TASK2K2P5.1 potassium channel TWIK-related acid-sensitive K(+) channel 2 TWIK-related acid-sensitive K+ channel 2; K2P5.1 potassium channel; Potassium channel subfamily K member 5; potassium channel, subfamily K, member 1 (TASK-2); potassium channel, subfamily, member 5 (KCNK5); potassium channel, two pore domain subfamily K, member 5; TWIK-related acid-sensitive K(+) channel 2; TWIK-related acid-sensitive K+ channel 2
Gene Aliases: K2p5.1; KCNK5; KCNK5b; TASK-2; TASK2
UniProt ID: (Human) O95279
Entrez Gene ID: (Human) 8645
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.