Invitrogen
Manufacturer Code:PA541045
Catalog # PIPA541045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human KCNK4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K2P4.1 potassium channel; Potassium channel subfamily K member 4; potassium channel two pore domain subfamily K member 4; potassium channel, subfamily K, member 4; potassium channel, two pore domain subfamily K, member 4; potassium inwardly-rectifying channel subfamily K member 4; potassium inwardly-rectifying channel, subfamily K, member 4; TRAAK; TWIK-related arachidonic acid-stimulated potassium channel protein; Two pore K(+) channel KT4.1; two pore K+ channel KT4.1; Two pore potassium channel KT4.1
Gene Aliases: K2p4.1; KCNK4; KT4.1; TRAAK; TRAAK1
UniProt ID: (Human) Q9NYG8, (Rat) G3V8V5
Entrez Gene ID: (Human) 50801, (Dog) 612120, (Horse) 100055490, (Pig) 100524785, (Guinea pig) 100732561, (Rat) 116489
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.