Invitrogen
Manufacturer Code:PA5109267
Catalog # PIPA5109267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide within human K2P4.1 aa 320-380. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K2p4.1 KCNK4 KT4.1 KT4.1b MLZ-622 Potassium channel subfamily K member 4 Tex40 TRAAK TRAAK1 TRAAKt TWIK-related arachidonic acid-stimulated potassium channel protein Two pore K(+) channel KT4.1 Two pore potassium channel KT4.1; K2P4.1 potassium channel; Potassium channel subfamily K member 4; potassium channel, subfamily K, member 4; potassium channel, two pore domain subfamily K, member 4; TRAAK; TWIK-related arachidonic acid-stimulated potassium channel protein; Two pore K(+) channel KT4.1; two pore K+ channel KT4.1; Two pore potassium channel KT4.1
Gene Aliases: K2p4.1; KCNK4; TRAAK; TRAAK1
UniProt ID: (Human) Q9NYG8
Entrez Gene ID: (Human) 50801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.