Proteintech
Manufacturer Code:271131AP150UL
Catalog # 50-173-5568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KCNK4 Fusion Protein Ag25931 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K2p4.1 KCNK4 TRAAK TRAAK1 Two pore K(+) channel KT4.1; K2P4.1 potassium channel; Potassium channel subfamily K member 4; potassium channel, subfamily K, member 4; potassium channel, two pore domain subfamily K, member 4; potassium inwardly-rectifying channel subfamily K member 4; potassium inwardly-rectifying channel, subfamily K, member 4; protein expressed in male leptotene and zygotene spermatocytes 622; TRAAK; TWIK-related arachidonic acid-stimulated potassium channel protein; Two pore K(+) channel KT4.1; two pore K+ channel KT4.1; Two pore potassium channel KT4.1
Gene Aliases: K2p4.1; KCNK4; KT4.1; MLZ-622; Tex40; TRAAK; TRAAK1; TRAAKt
UniProt ID: (Human) Q9NYG8, (Mouse) O88454, (Rat) G3V8V5
Entrez Gene ID: (Human) 50801, (Mouse) 16528, (Rat) 116489
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.