Novus Biologicals
Manufacturer Code:NBP242202UV
Catalog # NBP242202UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S374-48 |
Immunogen | Fusion protein amino acids 251-411 (cytoplasmic C-terminus) of rat Acid-sensitive potassium channel protein TASK or TASK1. Mouse: 96% identity (156/161 amino acids identical). Human: 76% identity (163/161 amino acids identical). <30% identity with TASK3. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: acid-sensitive potassium channel protein TASK; Acid-sensitive potassium channel protein TASK-1; Acid-sensitive potassium channel protein TASK-1 cardiac potassium channel K2p3.1 OAT1 potassium channel subfamily K member 3 potassium channel subfamily K member 3 potassium inwardly-rectifying channel subfamily K member 3 TASK1 TASK-1 TASKacid-sensitive potassium channel protein TASK TBAK1 TWIK-related acid-sensitive K(+) channel 1 TWIK-related acid-sensitive K+ channel two P domain potassium channel Two pore K(+) channel KT3.1 Two pore potassium channel KT3.1; cardiac potassium channel; Potassium channel subfamily K member 3; potassium channel, two pore domain subfamily K, member 3; potassium inwardly-rectifying channel, subfamily K, member 3; TWIK-related acid-sensitive K(+) channel 1; TWIK-related acid-sensitive K+ channel; two P domain potassium channel; Two pore K(+) channel KT3.1; Two pore potassium channel KT3.1
Gene Aliases: K2p3.1; KCNK3; OAT1; PPH4; TASK; TASK-1; TASK1; TBAK1
UniProt ID: (Human) O14649
Entrez Gene ID: (Human) 3777
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.