Invitrogen
Manufacturer Code:PA577589
Catalog # PIPA577589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)EDEKRDAEHRALLTRNGQ corresponding to amino acid residues 252-269 of human K2P 3.1 (TASK-1) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid-sensitive potassium channel protein TASK; Acid-sensitive potassium channel protein TASK-1; cardiac potassium channel; Potassium channel subfamily K member 3; potassium channel, subfamily K, member 3; potassium channel, two pore domain subfamily K, member 3; potassium inwardly-rectifying channel, subfamily K, member 3; TASK-1 Kcnk3; TWIK-related acid-sensitive K(+) channel 1; TWIK-related acid-sensitive K+ channel; two P domain potassium channel; Two pore K(+) channel KT3.1; Two pore potassium channel KT3.1
Gene Aliases: K2p3.1; KCNK3; OAT1; PPH4; rTASK; TASK; TASK-1; TASK1; TBAK1
UniProt ID: (Human) O14649, (Rat) O54912
Entrez Gene ID: (Human) 3777, (Rat) 29553
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.