Novus Biologicals
Manufacturer Code:NBP2841080.1ML
Catalog # NB032036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human KCNK3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid-sensitive potassium channel protein TASK; Acid-sensitive potassium channel protein TASK-1; Acid-sensitive potassium channel protein TASK-1 cardiac potassium channel K2p3.1 OAT1 potassium channel subfamily K member 3 potassium channel subfamily K member 3 potassium inwardly-rectifying channel subfamily K member 3 TASK1 TASK-1 TASKacid-sensitive potassium channel protein TASK TBAK1 TWIK-related acid-sensitive K(+) channel 1 TWIK-related acid-sensitive K+ channel two P domain potassium channel Two pore K(+) channel KT3.1 Two pore potassium channel KT3.1; cardiac potassium channel; Potassium channel subfamily K member 3; potassium channel, two pore domain subfamily K, member 3; potassium inwardly-rectifying channel, subfamily K, member 3; TWIK-related acid-sensitive K(+) channel 1; TWIK-related acid-sensitive K+ channel; two P domain potassium channel; Two pore K(+) channel KT3.1; Two pore potassium channel KT3.1
Gene Aliases: K2p3.1; KCNK3; OAT1; PPH4; TASK; TASK-1; TASK1; TBAK1
UniProt ID: (Human) O14649
Entrez Gene ID: (Human) 3777
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.