Invitrogen
Manufacturer Code:PA5114332
Catalog # PIPA5114332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to N terminal of Human KCNK2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A430027H14Rik AI848635 arachidonic acid sensitive tandem pore domain potassium channel EMBL:AAU06141.1 ECO:0000312 Ensembl:ENSRNOP00000003684 ECO:0000312 hTREK-1c hTREK-1e ion transport membrane protein K2p2.1 K2P2.1 potassium channel KCNK2 kcnk2 {ECO:0000312 Kcnk2 potassium channel outward rectifying potassium channel protein TREK-1 outward rectifying potassium channel protein TREK-1 {ECO:0000250 Potassium channel subfamily K member 2 potassium channel subfamily K member 2 {ECO:0000250 potassium channel subfamily k member 2 variant 1 potassium channel subfamily k member 2 variant 2 potassium channel subfamily K member 2 potassium channel two pore domain subfamily K member 2 potassium inwardly-rectifying channel subfamily K member 2 potassium two pore domain channel subfamily K member 2 PubMed:16248991} PubMed:24196565} RGD:621448} rTREK1d Stretch-activated potassium channel TREK-1 stretch-activated potassium channel TREK-1 {ECO:0000303 tandem-pore-domain potassium channel TREK-1 TPKC1 TREK trek 1 TREK1 Trek-1 TREK-1 K(+) channel subunit TREK-1 K(+) channel subunit {ECO:0000250 TWIK-related potassium channel 1 two pore domain potassium channel TREK-1 two pore domain potassium channel TREK-1 {ECO:0000303 Two pore potassium channel TPKC1 two pore potassium channel TPKC1 {ECO:0000250 two-pore potassium channel 1 UniProtKB:O95069}; arachidonic acid sensitive tandem pore domain potassium channel; ion transport membrane protein; K2P2.1 potassium channel; kcnk2 {ECO:0000312|EMBL:AAU06141.1, ECO:0000312|Ensembl:ENSRNOP00000003684, ECO:0000312|RGD:621448}; Outward rectifying potassium channel protein TREK-1; outward rectifying potassium channel protein TREK-1 {ECO:0000250|UniProtKB:O95069}; Potassium channel subfamily K member 2; potassium channel subfamily k member 2 variant 1; potassium channel subfamily k member 2 variant 2; potassium channel subfamily K member 2 {ECO:0000250|UniProtKB:O95069}; potassium channel, subfamily K, member 2; potassium channel, two pore domain subfamily K, member 2; potassium inwardly-rectifying channel, subfamily K, member 2; Stretch-activated potassium channel TREK-1; stretch-activated potassium channel TREK-1 {ECO:0000303|PubMed:16248991}; tandem-pore-domain potassium channel TREK-1; TREK-1 K(+) channel subunit; TREK-1 K(+) channel subunit {ECO:0000250|UniProtKB:O95069}; TWIK-related potassium channel 1; Two pore domain potassium channel TREK-1; two pore domain potassium channel TREK-1 {ECO:0000303|PubMed:24196565}; Two pore potassium channel TPKC1; two pore potassium channel TPKC1 {ECO:0000250|UniProtKB:O95069}; two-pore potassium channel 1
Gene Aliases: A430027H14Rik; AI848635; hTREK-1c; hTREK-1e; K2p2.1; KCNK2; rTREK1d; TPKC1; TREK; TREK-1; TREK1
UniProt ID: (Human) O95069, (Rat) Q920B6, (Mouse) P97438
Entrez Gene ID: (Human) 3776, (Rat) 170899, (Mouse) 16526
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.