Abnova
Manufacturer Code:PAB9063
Catalog # 89-134-131
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 417 of mouse Kcnk2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Kcnk2.
For Research Use Only
Protein Aliases: A430027H14Rik; AI848635; Outward rectifying potassium channel protein TREK-1; Potassium channel subfamily K member 2; TREK-1; TREK-1 K(+) channel subunit; Two pore potassium channel TPKC1
Gene Aliases: A430027H14Rik; AI848635; Kcnk2; TREK-1
UniProt ID: (Mouse) P97438
Entrez Gene ID: (Mouse) 16526
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.