Invitrogen
Manufacturer Code:PA116981
Catalog # 01-670-158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide within the N-terminal region (residues 1-100) of human TREK 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K2P2.1 potassium channel; Kcnk2 potassium channel; Outward rectifying potassium channel protein TREK-1; Potassium channel subfamily K member 2; potassium channel subfamily k member 2 variant 1; potassium channel subfamily k member 2 variant 2; potassium channel, subfamily K, member 2; potassium channel, two pore domain subfamily K, member 2; potassium inwardly-rectifying channel, subfamily K, member 2; tandem-pore-domain potassium channel TREK-1; TREK-1 K(+) channel subunit; TWIK-related potassium channel 1; Two pore domain potassium channel TREK-1; Two pore potassium channel TPKC1; Two-pore domain potassium channel TREK-1 antibody Potassium channel subfamily K member 2 antibody Twik-Related K+ Channel 1 antibody Kcnk2 antibo; two-pore potassium channel 1
Gene Aliases: A430027H14Rik; AI848635; hTREK-1c; hTREK-1e; K2p2.1; KCNK2; TPKC1; TREK; TREK-1; TREK1
UniProt ID: (Human) O95069, (Mouse) P97438
Entrez Gene ID: (Bovine) 282590, (Human) 3776, (Mouse) 16526
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.