Invitrogen
Manufacturer Code:PA577636
Catalog # PIPA577636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)EAEENPELKKFLDD corresponding to amino acid residues 62-75 of mouse K2P 18.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2P K ion channel TRESK; KCNK18; Potassium channel subfamily K member 18; potassium channel, subfamily K, member 18; tandem-pore K+ (K(2P)) channel; Tresk-2; TWIK-related individual K+ channel; TWIK-related spinal cord K+ channel; Two-pore-domain potassium channel TRESK
Gene Aliases: Gm781; Kcnk18; Tresk; Tresk-2; Tresk2; Trik
UniProt ID: (Rat) Q6Q1P3, (Mouse) Q6VV64
Entrez Gene ID: (Rat) 445371, (Mouse) 332396
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.