Invitrogen
Manufacturer Code:PA560810
Catalog # PIPA560810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KCNK18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K2p18.1 MGR13 TRESK TRESK-2 TRESK2 TRIK TWIK-related individual K+ channel TWIK-related individual potassium channel TWIK-related spinal cord K+ channel TWIK-related spinal cord potassium channel potassium channel subfamily K member 18; Potassium channel subfamily K member 18; potassium channel, subfamily K, member 18; potassium channel, two pore domain subfamily K, member 18; TWIK-related individual K+ channel; TWIK-related individual potassium channel; TWIK-related spinal cord K+ channel; TWIK-related spinal cord potassium channel
Gene Aliases: K2p18.1; KCNK18; MGR13; TRESK; TRESK-2; TRESK2; TRIK
UniProt ID: (Human) Q7Z418
Entrez Gene ID: (Human) 338567
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.