Invitrogen
Manufacturer Code:PA5114308
Catalog # PIPA5114308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide within mouse KCNK18 aa 1-150. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2P K ion channel TRESK; 2P K ion channel TRESK Gm781 K2p18.1 Kcnk18 mgR13 Potassium channel subfamily K member 18 potassium channel subfamily K member 18 potassium channel two pore domain subfamily K member 18 potassium two pore domain channel subfamily K member 18 tandem-pore K+ (K(2P)) channel TRESK Tresk2 TRESK-2 Trik TWIK-related individual K+ channel TWIK-related individual potassium channel TWIK-related spinal cord K+ channel TWIK-related spinal cord potassium channel Two-pore-domain potassium channel TRESK; Potassium channel subfamily K member 18; potassium channel, subfamily K, member 18; potassium channel, two pore domain subfamily K, member 18; tandem-pore K+ (K(2P)) channel; Tresk-2; TWIK-related individual K+ channel; TWIK-related individual potassium channel; TWIK-related spinal cord K+ channel; TWIK-related spinal cord potassium channel; Two-pore-domain potassium channel TRESK
Gene Aliases: Gm781; K2p18.1; KCNK18; MGR13; TRESK; TRESK-2; TRESK2; TRIK
UniProt ID: (Human) Q7Z418, (Rat) Q6Q1P3, (Mouse) Q6VV64
Entrez Gene ID: (Human) 338567, (Rat) 445371, (Mouse) 332396
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.