Abnova
Manufacturer Code:PAB19172
Catalog # 89-120-699
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 17 amino acids near center region of human KCNK18. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of KCNK18.
For Research Use Only
Protein Aliases: K2p18.1; Potassium channel subfamily K member 18; potassium channel, subfamily K, member 18; potassium channel, two pore domain subfamily K, member 18; TRESK; TRESK-2; TRESK2; TRIK; TWIK-related individual K+ channel; TWIK-related individual potassium channel; TWIK-related spinal cord K+ channel; TWIK-related spinal cord potassium channel
Gene Aliases: K2p18.1; KCNK18; MGR13; TRESK; TRESK-2; TRESK2; TRIK
UniProt ID: (Human) Q7Z418
Entrez Gene ID: (Human) 338567
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.