Invitrogen
Manufacturer Code:PA564188
Catalog # PIPA564188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KCNK15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid-sensitive potassium channel protein TASK-5; K2p15.1 KCNK11 KCNK14 KT3.3 TASK-5 TASK5 dJ781B1.1 TWIK-related acid-sensitive K(+) channel 5 acid-sensitive potassium channel protein TASK-5 potassium channel subfamily K member 15 potassium channel subfamily K member 14 two pore K(+) channel KT3.3 two pore potassium channel KT3.3; Potassium channel subfamily K member 15; potassium channel, subfamily K, member 14; potassium channel, two pore domain subfamily K, member 15; TWIK-related acid-sensitive K(+) channel 5; Two pore K(+) channel KT3.3; Two pore potassium channel KT3.3
Gene Aliases: dJ781B1.1; K2p15.1; KCNK11; KCNK14; KCNK15; KT3.3; TASK-5; TASK5
UniProt ID: (Human) Q9H427
Entrez Gene ID: (Human) 60598
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.