Invitrogen
Manufacturer Code:PA5101759
Catalog # PIPA5101759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KCNK15 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid-sensitive potassium channel protein TASK-5; Acid-sensitive potassium channel protein TASK-5 dJ781B1.1 K2p15.1 KCNK11 KCNK14 Kcnk15 KT3.3 Potassium channel subfamily K member 15 potassium channel subfamily K member 15 (TASK-5) potassium channel subfamily K member 14 potassium channel subfamily K member 15 potassium channel subfamily K member 15 (TASK-5) potassium channel two pore domain subfamily K member 15 potassium two pore domain channel subfamily K member 15 rTASK-5 Task5 TASK-5 TWIK-related acid-sensitive K(+) channel 5 two pore K(+) channel KT3.3 Two pore potassium channel KT3.3; Potassium channel subfamily K member 15; potassium channel subfamily K member 15 (TASK-5); potassium channel, subfamily K, member 14; potassium channel, subfamily K, member 15 (TASK-5); potassium channel, two pore domain subfamily K, member 15; rTASK-5; TWIK-related acid-sensitive K(+) channel 5; Two pore K(+) channel KT3.3; Two pore potassium channel KT3.3
Gene Aliases: dJ781B1.1; K2p15.1; KCNK11; KCNK14; KCNK15; KT3.3; TASK-5; TASK5
UniProt ID: (Human) Q9H427, (Rat) Q8R5I0
Entrez Gene ID: (Human) 60598, (Rat) 156873
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.