Invitrogen
Manufacturer Code:PA577635
Catalog # PIPA577635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)EERLANFSRGHNLSRE corresponding to amino acid residues 54-69 of rat K2P 13.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KCNK13; Potassium channel subfamily K member 13; potassium channel, subfamily K, member 13; prdx1; Tandem pore domain halothane-inhibited potassium channel 1; tandem pore domain potassium channel THIK-1; THIK-1
Gene Aliases: BB085247; F730021E22Rik; Gm1570; Gm1685; Kcnk13; prdx1; THIK-1
UniProt ID: (Mouse) Q8R1P5, (Rat) Q9ERS0
Entrez Gene ID: (Mouse) 217826, (Rat) 64120
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.