Invitrogen
Manufacturer Code:PA572686
Catalog # PIPA572686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KCNK13 antibody for a 15 amino acid synthetic peptide near the center of human KCNK13. The immunogen is within amino acids 150 - 200 of KCNK13. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K2P13.1 potassium channel; Potassium channel subfamily K member 13; potassium channel, subfamily K, member 13; potassium channel, two pore domain subfamily K, member 13; prdx1; Tandem pore domain halothane-inhibited potassium channel 1; tandem pore domain potassium channel THIK-1; THIK-1; THIK1 THIK-1 K2p13.1
Gene Aliases: BB085247; F730021E22Rik; Gm1570; Gm1685; K2p13.1; KCNK13; prdx1; THIK-1; THIK1
UniProt ID: (Human) Q9HB14, (Mouse) Q8R1P5, (Rat) Q9ERS0
Entrez Gene ID: (Human) 56659, (Mouse) 217826, (Rat) 64120
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.