Abnova
Manufacturer Code:PAB25687
Catalog # 89-127-308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 14 amino acids at C-terminus of human KCNK12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of KCNK12.
For Research Use Only
Protein Aliases: Potassium channel subfamily K member 12; potassium channel, subfamily K, member 12; potassium channel, two pore domain subfamily K, member 12; Tandem pore domain halothane-inhibited potassium channel 2; tandem pore domain potassium channel THIK-2; THIK-2; THIK2
Gene Aliases: K2p12.1; KCNK12; THIK-2; THIK2
UniProt ID: (Human) Q9HB15
Entrez Gene ID: (Human) 56660
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.