Abnova
Manufacturer Code:H00056660A01
Catalog # 89-107-142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 166 a.a. &bsim 215 a.a) partial recombinant protein with GST tag.,KCNK12 (NP_071338 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant KCNK12.
For Research Use Only
Protein Aliases: Potassium channel subfamily K member 12; potassium channel, subfamily K, member 12; potassium channel, two pore domain subfamily K, member 12; Tandem pore domain halothane-inhibited potassium channel 2; tandem pore domain potassium channel THIK-2; THIK-2; THIK2
Gene Aliases: K2p12.1; KCNK12; THIK-2; THIK2
UniProt ID: (Human) Q9HB15
Entrez Gene ID: (Human) 56660
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.