Invitrogen
Manufacturer Code:PA541010
Catalog # PIPA541010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KCNK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: double-pore potassium channel; Inward rectifying potassium channel protein TWIK-1; KCNK1; kcnk1 {ECO:0000312|RGD:621447}; Potassium channel K2P1; Potassium channel KCNO1; Potassium channel subfamily K member 1; Potassium channel subfamily K member 1 DPK HOHO K2p1.1 TWIK-1 TWIK1 K2P1 KCNO1; potassium channel TWIK-1; potassium channel, subfamily K, member 1; potassium channel, two pore domain subfamily K, member 1; potassium inwardly-rectifying channel, subfamily K, member 1; putative potassium channel TWIK; rTWIK; tandem of P domains in a weak inward rectifying K+ channel 1; two-pore potassium channel
Gene Aliases: AI788889; DPK; HOHO; HOHO1; K2P1; K2p1.1; KCNK1; KCNO1; rabKCNK1; Twik; TWIK-1; TWIK1
UniProt ID: (Rabbit) Q5UE96, (Bovine) Q0P5A0, (Human) O00180, (Rat) Q9Z2T2, (Guinea pig) Q8R454, (Mouse) O08581
Entrez Gene ID: (Pig) 100156432, (Rabbit) 100008962, (Horse) 100059489, (Bovine) 505563, (Dog) 488965, (Human) 3775, (Rat) 59324, (Guinea pig) 100135567, (Mouse) 16525
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.