Novus Biologicals
Manufacturer Code:NBP297436DL594
Catalog # NB043459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human KCNK1 fragment. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DPK HOHO1 Inward rectifying potassium channel protein TWIK-1 K2P1 K2p1.1 KCNO1 Potassium channel KCNO1 potassium channel subfamily K member 1 potassium channel subfamily K member 1 potassium inwardly-rectifying channel subfamily K member 1 TWIK-1 TWIK1HOHO; Inward rectifying potassium channel protein TWIK-1; Potassium channel K2P1; Potassium channel KCNO1; Potassium channel subfamily K member 1; potassium channel, two pore domain subfamily K, member 1; potassium inwardly-rectifying channel, subfamily K, member 1; tandem of P domains in a weak inward rectifying K+ channel 1
Gene Aliases: DPK; HOHO; HOHO1; K2P1; K2p1.1; KCNK1; KCNO1; TWIK-1; TWIK1
UniProt ID: (Human) O00180
Entrez Gene ID: (Human) 3775
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.