Novus Biologicals
Manufacturer Code:NBP2831040.1ML
Catalog # NB031033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of rat KCNJ14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-sensitive inward rectifier potassium channel 14; ATP-sensitive inward rectifier potassium channel 14 IRK4 KIR2.4 potassium inwardly-rectifying channel subfamily J member 14; Inward rectifier K(+) channel Kir2.4; inwardly rectifying potassium channel KIR2.4; IRK-4; Potassium channel, inwardly rectifying subfamily J member 14; potassium channel, inwardly rectifying subfamily J, member 14; potassium inwardly-rectifying channel, subfamily J, member 14
Gene Aliases: IRK4; KCNJ14; KIR2.4
UniProt ID: (Human) Q9UNX9
Entrez Gene ID: (Human) 3770
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.