Invitrogen
Manufacturer Code:PA577647
Catalog # PIPA577647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)SLQTKQRRPSKD corresponding to amino acid residues 10-21 of rat KChIP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A-type potassium channel modulatory protein 1; A-type potassium channel modulatory protein 1.2; KChIP1; KCNIP1; Kv channel interacting protein 1; Kv channel-interacting protein 1; potassium channel interacting protein 1; Potassium channel-interacting protein 1; Vesicle APC-binding protein
Gene Aliases: KCHIP1; Kchip1.2; KCNIP1; VABP
UniProt ID: (Human) Q9NZI2, (Mouse) Q9JJ57, (Rat) Q8R426
Entrez Gene ID: (Human) 30820, (Mouse) 70357, (Rat) 65023
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.