Invitrogen
Manufacturer Code:PA541041
Catalog # PIPA541041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human KCNIP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A-type potassium channel modulatory protein 1; A-type potassium channel modulatory protein 1.2; KChIP1; Kv channel interacting protein 1; Kv channel interacting protein 1 VABP KCHIP1; Kv channel-interacting protein 1; potassium channel interacting protein 1; Potassium channel-interacting protein 1; Vesicle APC-binding protein
Gene Aliases: KCHIP1; Kchip1.2; KCNIP1; VABP
UniProt ID: (Human) Q9NZI2, (Mouse) Q9JJ57, (Rat) Q8R426
Entrez Gene ID: (Bovine) 541458, (Human) 30820, (Rabbit) 100339714, (Dog) 489125, (Horse) 100059195, (Mouse) 70357, (Rat) 65023
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.