Invitrogen
Manufacturer Code:PA568691
Catalog # PIPA568691
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human KCNH8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ELK channel 3; ELK ELK1 elk3 Kv12.1; ELK1; ether-a-go-go-like potassium channel 1; Ether-a-go-go-like potassium channel 3; hElk1; potassium channel, voltage gated eag related subfamily H, member 8; Potassium voltage-gated channel subfamily H member 8; potassium voltage-gated channel, subfamily H (eag-related), member 8; potassium voltage-gated channel, subfamily H, member 8; Voltage-gated potassium channel subunit Kv12.1
Gene Aliases: C130090D05Rik; ELK; ELK1; ELK3; Ets-1; KCNH8; Kv12.1
UniProt ID: (Human) Q96L42, (Mouse) P59111, (Rat) Q9QWS8
Entrez Gene ID: (Rabbit) 100349166, (Dog) 485649, (Human) 131096, (Pig) 100626552, (Horse) 100052056, (Mouse) 211468, (Rat) 246325
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.