Invitrogen
Manufacturer Code:PA532079
Catalog # PIPA532079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 264 of Human KCNH7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eag-related protein 3; ERG-3; ERG3 HERG3 Kv11.3 ERG-3 eag related protein 3 eag-related protein 3 ether-a-go-go related gene potassium channel 3 ether-a-go-go-related gene potassium channel 3 ether-a-go-go-related protein 3 potassium channel subunit HERG-3 potassium voltage-gated channel subfamily H me; Ether-a-go-go-related gene potassium channel 3; ether-a-go-go-related protein 3; potassium channel subunit HERG-3; potassium channel, voltage gated eag related subfamily H, member 7; Potassium voltage-gated channel subfamily H member 7; Voltage-gated potassium channel subunit Kv11.3
Gene Aliases: ERG3; HERG3; KCNH7; Kv11.3
UniProt ID: (Human) Q9NS40
Entrez Gene ID: (Human) 90134
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.