Proteintech
Manufacturer Code:136221AP150UL
Catalog # 50-172-8883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KCNH7 Fusion Protein Ag4532 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Eag related protein 3 ERG 3 ERG3 hERG 3 HERG3 KCNH7 Kv11.3; eag-related protein 3; ERG-3; Ether-a-go-go-related gene potassium channel 3; ether-a-go-go-related protein 3; potassium channel erg3; potassium channel protein erg3; potassium channel subunit HERG-3; potassium channel, voltage gated eag related subfamily H, member 7; Potassium voltage-gated channel subfamily H member 7; potassium voltage-gated channel, subfamily H (eag-related), member 7; Voltage-gated potassium channel subunit Kv11.3
Gene Aliases: 9330137I11Rik; ERG3; HERG3; KCNH7; Kv11.3
UniProt ID: (Human) Q9NS40, (Mouse) Q9ER47, (Rat) O54852
Entrez Gene ID: (Human) 90134, (Mouse) 170738, (Rat) 170739
Molecular Function:
cyclic nucleotide-gated ion channel
ion channel
ligand-gated ion channel
potassium channel
receptor
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.