Invitrogen
Manufacturer Code:PA577629
Catalog # PIPA577629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide TLNFVEFNLEKHRS(C) corresponding to amino acid residues 185-198 of human Kv 11.2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Eag-related gene member 2; eag-related protein 2; ERG-2; Ether-a-go-go-related gene potassium channel 2; ether-a-go-go-related protein 2; KCNH6; potassium channel, voltage gated eag related subfamily H, member 6; Potassium voltage-gated channel subfamily H member 6; potassium voltage-gated channel, subfamily H (eag-related), member 6; Voltage-gated potassium channel subunit Kv11.2
Gene Aliases: ERG-2; ERG2; hERG-2; HERG2; KCNH6; Kv11.2; m-erg2
UniProt ID: (Human) Q9H252, (Rat) O54853
Entrez Gene ID: (Human) 81033, (Rat) 116745, (Mouse) 192775
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.