Invitrogen
Manufacturer Code:PA541061
Catalog # PIPA541061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human KCNH6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Eag-related gene member 2; eag-related protein 2; ERG-2; Ether-a-go-go-related gene potassium channel 2; ether-a-go-go-related protein 2; potassium channel, voltage gated eag related subfamily H, member 6; Potassium voltage-gated channel subfamily H (eag-related) member 6 ERG2 HERG2 Kv11.2; Potassium voltage-gated channel subfamily H member 6; potassium voltage-gated channel, subfamily H (eag-related), member 6; Voltage-gated potassium channel subunit Kv11.2
Gene Aliases: ERG-2; ERG2; hERG-2; HERG2; KCNH6; Kv11.2; m-erg2
UniProt ID: (Human) Q9H252, (Rat) O54853
Entrez Gene ID: (Bovine) 527872, (Horse) 100054322, (Rabbit) 100355076, (Human) 81033, (Dog) 100856219, (Rat) 116745, (Mouse) 192775
Molecular Function:
cyclic nucleotide-gated ion channel
ion channel
ligand-gated ion channel
potassium channel
receptor
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.