Abnova
Manufacturer Code:PAB22454
Catalog # 89-124-163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human KCNH5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant KCNH5.
For Research Use Only
Protein Aliases: EAG2; Ether-a-go-go potassium channel 2; ether-a-go-go-related potassium channel 2; H-EAG2; hEAG2; Kv10.2; potassium channel, voltage gated eag related subfamily H, member 5; Potassium voltage-gated channel subfamily H member 5; potassium voltage-gated channel, subfamily H (eag-related), member 5; Voltage-gated potassium channel subunit Kv10.2
Gene Aliases: EAG2; H-EAG2; hEAG2; KCNH5; Kv10.2
UniProt ID: (Human) Q8NCM2
Entrez Gene ID: (Human) 27133
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.