Invitrogen
Manufacturer Code:PA577625
Catalog # PIPA577625
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide AFLLKETEEGPPATEC corresponding to residues 430-445 of human KV 11.1 (HERG) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Eag homolog; eag-related protein 1; ERG-1; Ether-a-go-go-related gene potassium channel 1; ether-a-go-go-related potassium channel protein; ether-a-go-go-related protein 1; Potassium Channel Kv11.1; potassium channel, voltage gated eag related subfamily H, member 2; Potassium voltage-gated channel subfamily H member 2; potassium voltage-gated channel, subfamily H (eag-related), member 2; r-ERG; RERG; Voltage-gated potassium channel subunit Kv11.1
Gene Aliases: ERG; ERG-1; ERG1; H-ERG; HERG; HERG1; KCNH2; Kv11.1; LQT2; SQT1
UniProt ID: (Human) Q12809, (Rat) O08962
Entrez Gene ID: (Human) 3757, (Rat) 117018
Molecular Function:
cyclic nucleotide-gated ion channel
ion channel
ligand-gated ion channel
potassium channel
receptor
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.