Invitrogen
Manufacturer Code:PA577621
Catalog # PIPA577621
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide GDPAKRKGWARFKDAC corresponding to amino acid residues 802-817 of rat KV 10.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAG channel 1; EAG1; ether a go-go; Ether-a-go-go potassium channel 1; ether-a-go-go, Drosophila, homolog of; KCNH1; potassium channel, voltage gated eag related subfamily H, member 1; Potassium voltage-gated channel subfamily H member 1; potassium voltage-gated channel, subfamily H (eag-related), member 1; r-eag; Voltage-gated potassium channel subunit Kv10.1
Gene Aliases: EAG; EAG1; h-eag; hEAG1; KCNH1; Kv10.1; M-eag; TMBTS; ZLS1
UniProt ID: (Human) O95259, (Rat) Q63472, (Mouse) Q60603
Entrez Gene ID: (Human) 3756, (Rat) 65198, (Mouse) 16510
Molecular Function:
cyclic nucleotide-gated ion channel
ion channel
ligand-gated ion channel
potassium channel
receptor
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.