Invitrogen
Manufacturer Code:PA575527
Catalog # PIPA575527
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 762-807 of Human KCNH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAG channel 1; EAG1; ether a go-go; Ether-a-go-go potassium channel 1; ether-a-go-go, Drosophila, homolog of; potassium channel, voltage gated eag related subfamily H, member 1; Potassium voltage-gated channel subfamily H member 1; Potassium voltage-gated channel subfamily H member 1 Ether-a-go-go potassium channel 1 EAG channel 1 h-eag hEAG1 Voltage-gated potassium channel subunit Kv10.1 KCNH1 EAG EAG1; potassium voltage-gated channel, subfamily H (eag-related), member 1; r-eag; Voltage-gated potassium channel subunit Kv10.1
Gene Aliases: EAG; EAG1; h-eag; hEAG1; KCNH1; Kv10.1; M-eag; TMBTS; ZLS1
UniProt ID: (Human) O95259, (Rat) Q63472, (Mouse) Q60603
Entrez Gene ID: (Human) 3756, (Rat) 65198, (Mouse) 16510
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.